Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ M-270 Epoxy

Catalog No. p-4924202
Encompass_Preferred
Click to view available options
Quantity:
60 mg
300 mg

Includes

60mg Dynabeads M-270 Epoxy, approx. 6.7 x 107 beads/mg, supplied as a freeze-dried powder.

Dynabeads™ M-270 Epoxy beads are 2.8 μm superparamagnetic beads containing surface epoxy groups.

Dynabeads™ M-270 Epoxy beads are 2.8 μm superparamagnetic beads containing surface epoxy groups. The beads covalently bind primary amino and sulfhydryl groups in proteins and peptides, making them ideal for coupling antibodies, peptides, intact proteins, and functional enzymes. These hydrophylic, neutral pH beads exhibit extremely low nonspecific binding of proteins and dyes, which reduces the need for blocking agents. Also, Dynabeads™ M-270 Epoxy Beads do not contain Tween™ detergent, and so are ideal for use in mass spectrometry (MS) analysis.

• Ideal for immunoprecipitation (IP) of proteins and protein complexes
• Gentle, yet rapid magnetic separation and short incubation times allow identification of transient and labile complexes
• Covalent antibody-coupling to the Dynabeads™ avoids co-elution of the antibody with the target protein

Adaptable for various applications
Fast surface binding kinetics and rapid magnetic separation characteristics make Dynabeads™ M-270 Epoxy beads the ideal choice for applications such as immunoprecipitation/purification of proteins and protein complexes, coupling functional enzymes to the bead surface for downstream assays, and identifying protein binding partners. Captured proteins and protein complexes are easily separated, washed, and eluted using a DynaMag™ magnet, and the magnetic separation properties of the Dynabeads™ .

Coupling procedure
Covalent coupling is performed overnight by incubating the desired ligand (commonly antibodies, peptides, or proteins) with Dynabeads™ M-270 Epoxy beads. The binding capacity varies depending on the ligand used, but is typically 5–10 μg IgG per milligram of beads. Coupling occurs at neutral pH and high salt concentration. The coupling reaction can be performed over a broad range of temperatures to match the stability characteristics of your ligand.

Upon completion of the ligand coupling step, the reactive moieties on the Dynabeads™ M-270 Epoxy beads are deactivated to prevent further coupling. Once the coupling reaction is completed, these nonporous, hydrophilic, pH neutral beads exhibit extremely low nonspecific binding.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 60 mg Dynabeads™ M-270 Epoxy, ∼6.7 x 107 beads/mg, supplied as a freeze-dried powder.

Store at 2°C to 8°C.
Diameter (Metric) 2.8 μm
Elution Volume Min 5 μL
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality Epoxy
Product Line DYNAL™, Dynabeads™
Quantity 60 mg
Type Epoxy Bead
Format Freeze Dried
Purity or Quality Grade Research Grade
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.