Learn More
Invitrogen™ Dynabeads™ Streptavidin Trial Kit

Description
Includes
1mL (10mg/mL) of each of the following bead-types: Dynabeads™ M-280 Streptavidin (2.8μm beads supplied at a concentration of 6-7x108 beads per ml, Dynabeads™ M-270 Streptavidin (2.8μm beads supplied at a concentration of 6-7x108 beads per mL, Dynabeads™ MyOne™ Streptavidin C1 (1μm beads supplied at a concentration of 7-12x109 beads per mL, Dynabeads™ MyOne™ Streptavidin T1 (1μm beads supplied at a concentration of 7-12x109 beads per mL
The Dynabeads Streptavidin Trial Kit contains four different streptavidin-coated Dynabeads products making it more convenient to test out which of the beads perform best in a specific application. The kit contains 1 mL each of Dynabeads M-280 Streptavidin, Dynabeads M-270 Streptavidin, Dynabeads MyOne Streptavidin C1, and Dynabeads MyOne Streptavidin T1. The streptavidin-coated Dynabeads magnetic beads are monosized (1 or 2.8 μm in diameter depending on the product), monodispersed, and superparamagnetic. The products are intended for the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands.
Benefits of Dynabeads Streptavidin Trial Kit include:
• Performance—high sensitivity and reproducibility secure consistent results
• Efficiency—fast binding kinetics and high binding capacity
• Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
• Automation ready—easy automation for high-throughput needs on the Kingfisher Purification System
Consistency and reproducibility
The monolayer of streptavidin leaves most of the biotin-binding sites available for the binding of free biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer help ensure negligible leakage, giving batch consistency and reproducibility of results.
Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications such as immunoassays, purification of proteins, cells, extracellular vesicles/exosomes, viruses, exosomes, or phage display and bio-panning.
Automation ready with the KingFisher Purification System
Dynabeads magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems or liquid handlers.
Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Streptavidin Trial Kit in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Bead Type | Magnetic Polystyrene |
Color | Brown |
Concentration | 10 mg/mL |
Content And Storage | The Dynabeads Streptavidin Trial Kit contains 1 mL (10 mg/mL) each of four different streptavidin-coated Dynabeads so you can find the best match for your application. • Dynabeads M-280 Streptavidin • Dynabeads M-270 Streptavidin • Dynabeads MyOne Streptavidin C1 • Dynabeads MyOne Streptavidin T1 Store at 2–8°C. |
Description | Recombinant streptavidin covalently coupled to Dynabeads |
Format | Beads in suspension |
Isolation Technology | Magnetic beads |
Diameter (Metric) | 1, 2.8 μm |
Sample Type | Any Sample Type |
For Use With (Equipment) | KingFisher™ Sample Purification System, DynaMag™ magnets |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.