Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Untouched™ Mouse CD4 Cells Kit

Catalog No. p-4924228
Encompass_Preferred
Click to view available options
Quantity:
4 mL
2 x 10 mL

Includes

20mL Mouse Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process approx. 1 x 109 cells

Dynabeads™ Untouched™ Mouse CD4 Cells contains both magnetic beads and an Antibody Mix to deplete all leucocytes from mouse spleen or lymph node cells, leaving only the target mouse CD4+ cells.

Dynabeads Untouched Mouse CD4 Cells supplies both 4.5μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix, together designed for the depletion of CD4-negative leucocytes from mouse spleen or lymph node single-cell suspensions. Following the depletion procedure, the negatively isolated, untouched target mouse CD4+ cells are pure, viable, and ready to be used in any downstream application.

  • Rapid isolation of untouched mouse CD4
  • T cells—no columns required
  • Pure and viable mouse CD4+ T cells are ready for any downstream application

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD4)
Content And Storage The Kit Contains: 4 mL Mouse Depletion Dynabeads™ and 0.4 mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119.
Store at 2°C to 8°C.
Product Type Cell Isolation Kit
Final Product Type Cells
Isolation Technology Negative Isolation
No. of Cells Processes 2 x 108 cells
Product Line DYNAL™, Dynabeads™
Purity or Quality Grade Research Grade
Quantity 4 mL
Reactivity Mouse
Sample Type Lymph Node, MNC, Spleen
Shipping Condition Room Temperature
Target Species Mouse
Diameter (Metric) 4.5 μm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.