Learn More
Invitrogen™ E-Gel™ 96 Agarose Gels

Description
Includes
Each box of E-Gel 96 gels contains 8 gels and a manual.
Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.
Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.
Each gel contains 96 sample lanes and 8 marker lanes, ideal for screening multiple PCR products, plasmid preparations, and restriction digests. The E-Gel 96 loading format is compatible with multi-channel pipettors and the most commonly used 8-, 12-, and 96-pin liquid handling robots. With a 12-minute run time, up to 20,000 samples can be resolved in a single day.
The unique 96-well staggered-well format provides a 1.6-cm run length with the following separation ranges for DNA fragments:
- 1% agarose gels: DNA fragments between 400 bp and 10 kb
- 2% agarose gels: DNA fragments between 100 bp and 2 kb
Each cassette is labeled with an unique EAN39 bar code that is recognized by most commercially available robotic bar code readers.
Features of E-Gel 96 gels include:
- High throughput analysis—contain 96 sample lanes and 8 marker lanes
- Automation compatible—suitable for use with multi-channel pipettors and commonly used 8-, 12-, and 96-pin liquid handling robots
- Scalable—resolve up to 384 samples in one run by combining up to four E-Base Mother and E-Base Daughter devices
- Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
- High performance—detect 3 ng of sample DNA with a separation time as short as 12–17 minutes
Safer option
Choose SYBR Safe DNA stained gels to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is safer for the user, safer for the environment, and can save institutional overhead costs for disposal.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Running and imaging devices
High-throughput E-Gel agarose gels require the E-Base Electrophoresis Device for gel running.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with or without SYBR Safe are available in 1% and 2% gel percentages.
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Content And Storage | Each box contains 8 gels and a manual
|
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 2000 bp |
Stain Type | Ethidium Bromide |
Wells | 96-well |
Quantity | 8 Gels |
Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.