Learn More
Invitrogen™ Taq DNA Polymerase, recombinant
Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.
Supplier: Invitrogen™ 10342053

Includes: Taq DNA Polymerase (5U/μL); 10X PCR buffer (200mM Tris-HCl pH 8.4, 500mM KCl); Magnesium Chloride (50mM)
Description
Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.
Features
- A choice of recombinant or native enzyme
- Amplification of PCR products up to 5 kb in size
- Enzyme is licensed and qualified for PCR
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- RT-PCR
- Sequencing ssDNA
- Cycle sequencing
Notes
- One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
- For superior PCR performance, DreamTaq DNA Polymerase is recommended.
Order Info
Shipping Condition: Approved for shipment on wet or dry ice

Specifications
5 U/μL | |
Stand-alone enzyme | |
qPCR, Standard PCR | |
Standard | |
DNA Polymerase | |
Dry Ice | |
1X | |
100 Reactions | |
Separate Components | |
DNA |
• Taq DNA Polymerase (5 U/μL), 20 μL • 10X PCR buffer, 1.5 mL • 50 mM MgCl2, 1 mL Store at -10°C to -30°C in a non-frost-free freezer. Guaranteed stable for 6 months when stored properly. |
|
Low | |
Taq DNA Polymerase | |
5' - 3' | |
100 Units | |
Standard PCR | |
No | |
3'-A | |
5 kb or less |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.