Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent

Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments

Supplier:  Invitrogen™ 13778100

Encompass_Preferred

 View more versions of this product

Catalog No. 13-778-100


Add to Cart

Includes: Contains one vial (0.75mL) Lipofectamine RNAiMAX Transfection Reagent.

Description

Description

Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.

With Lipofectamine™ RNAiMAX Transfection Reagent you will get:

  • Superior transfection efficiency requiring lower RNAi concentrations, leading to more effective gene knockdown with minimal nonspecific effects
  • Easy optimization due to minimal cytotoxicity across a 10-fold concentration range of transfection reagent
  • Superior transfection efficiencies for miRNA antagonists and mimics
  • Compatibility with a broad range of cell types, providing the most versatile approach to all of your gene silencing experiments
  • A simple and rapid protocol for consistent and reproducible results

    High knockdown in a wide range of cells
    Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.

    Simple, high-throughput–ready transfections
    Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.

    Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.

    Order Info

    Shipping Condition: Qualified for Ambient or Wet Ice

    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Each unit contains one vial of reagent. Store at 2–8°C.
    6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
    Lipid-based Transfection
    Sustainable packaging
    Lipofectamine™
    0.1 mL
    Approved for shipment at Room Temperature or on Wet Ice
    Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    siRNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR), RNAi, miRNA
    Transfection
    High-throughput Compatible
    Transfection Reagent
    Yes
    Product Suggestions

    Product Suggestions

    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit

    For Research Use Only. Not for use in diagnostic procedures.