Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 24-Well Magnetic Separator

Catalog No. cs15024
Encompass_Preferred
Click to view available options
Quantity:
1 Ea.

Includes

One Magnetic Separator

Contains six neodymium magnets that aligned with wells of a 24-well plate for efficient processing of DNA bound to magnetic beads.

  • Contains six neodymium magnets that aligned with wells of a 24-well plate for efficient processing of DNA bound to magnetic beads
  • GeneCatcher technology is novel magnetic bead-based method that reliably and efficiently purifying DNA without any centrifugation steps
  • GeneCatcher gDNA Blood Kits facilitate efficient and reliable extraction of genomic DNA (gDNA) from samples of human blood, even when the samples have been archived, poorly stored, or collected in the presence of anticoagulants such as EDTA, heparin, or citrate

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one Magnetic Separator, which may be stored at room temperature.
For Use With (Equipment) MagnaRack™ Magnetic Separator, Magna-Sep™ Magnetic Particle Separator
Shipping Condition Room Temperature
Product Line MagnaRack™
Quantity 1 Ea.
Type Magnetic Separator

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.