Learn More
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit With ROX Non-distribution product as customer accommodation.
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
Manufacturer:  Invitrogen™ 11745100
 View more versions of this product
Catalog No. 11-745-100
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes: SuperScript™ III/Platinum™ Taq Mix, 2X Reaction Mix with ROX and 1mL Magnesium Sulfate (50mM). Guaranteed stable for 6 months when properly stored.
Specifications
Description
Two kit configurations allow your choice of detection methods:
  • SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
  • SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection for use with LUX™ fluorogenic primers or dual-labeled probes (e.g. TaqMan™)
Order Info
Dry Ice
Description & Specifications
Specifications
Primer-Probe Detection
7000 System, 7300 System, 7700 System, 7900HT System
DNA
Store all components at -20°C. Store ROX Reference dye in the dark.
100 Reactions
Real time PCR (qPCR), real time PCR-based gene expression profiling
Includes SuperScript™ III RT/Platinum™ Taq Mix, 2X Reaction Mix (containing SYBR™ Green I, ROX Reference Dye, dNTPs, and MgSO4), and 50mM MgSO4. SuperScript™ III Platinum™ One-Step qRT-PCR Kit with ROX includes SuperScript™ III RT/Platinum™ Taq Mix, 2X Reaction Mix (containing ROX Reference Dye, dNTPs and MgSO4), and 50mM MgSO4.
Standard
100 Reactions