Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG

Catalog No. p-4927007
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-733-046 500 Reactions
11-733-038 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-733-046 Supplier Invitrogen™ Supplier No. 11733046
Only null left
Add to Cart
Edge
Add to Cart

Includes

Platinum SYBR Green qPCR SuperMix-UDG (2 × 1.25mL), 50mM Magnesium Chloride (1mL), ROX Reference Dye (100μL), 20X BSA (300μL)

Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.

The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.

  • Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
  • 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
  • Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
  • SYBR™ Green I dye for simple and easy detection of real-time product

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Platinum™ SYBR™ Green qPCR SuperMix-UDG (12.5 mL)
• 50 mM Magnesium Chloride (2 × 1 mL)
• ROX Reference Dye (500 μL)
• 20X BSA (1.3 mL)

Sufficient reagents are provided for 500 reactions of 50 μL each. Stored at -20°C.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ
Product Line Platinum, SYBR
Product Type Real Time PCR SYBR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions of 50 μL Each
For Use With (Application) Gene Expression
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 12.5 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.