Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix Universal

Catalog No. p-4927011
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-762-02K 2000 Reactions
11-762-100 100 Reactions
11-762-500 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-762-02K Supplier Invitrogen™ Supplier No. 1176202K
Only null left
Add to Cart
Add to Cart
Request a Quote

Includes

2X SYBR GreenER qPCR SuperMix and a separate tube of ROX reference dye

Incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms including ABI 7500, Corbett Rotor-Gene™ etc.

SYBR™ GreenER™ qPCR SuperMixes Universal incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms, including ABI 7500, Corbett Rotor-Gene™, MJ Chromo4™ and Opticon™, and Stratagene instruments. The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with. A novel dsDNA-binding dye exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings. An optimized buffer system improves sensitivity and provides excellent long-term stability. Uracil DNA glycosylase reduces carryover contamination in qPCR.

Eight cDNA targets and eight genomic DNA targets were amplified from 1ng of cDNA and 6pg of gDNA, respectively, on the ABI PRISM™ 7900HT Sequence Detection System using the SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments. For all 16 targets, detection was positive in the presence of template but negative for all no-template controls.

  • The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range

Consistent Specificity and Low-Copy Detection:
  • The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets
  • The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity
  • With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage SYBR™ GreenER™ qPCR SuperMix Universal contains:

2X SYBR™ GreenER™ qPCR SuperMix and a separate tube of ROX reference dye. Store at +4°C upon receipt. Guaranteed stable for 6 months when properly stored.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P, MJ Chromo4, MJ Opticon
Product Line Platinum, SYBR GreenER
Product Type Real Time PCR SYBR Master Mix
Quantity 2,000 reactions
Sufficient For 2000 Reactions
For Use With (Application) Gene Expression
No. of Reactions 2000 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less
What is the difference in sensitivity between TaqMan chemistry vs. SYBR Green reagent chemistry?

Sensitivity can actually be equivalent when using TaqMan chemistry and SYBR Green reagent chemistry. It might seem that a TaqMan assay with fluorescent signal generated by a sequence-specific probe would always be more sensitive than a SYBR Green reagent assay, but a poorly designed TaqMan assay could theoretically be less specific than a well-designed SYBR Green reagent assay. However, the potential for detection of primer dimers and non-specific products using SYBR Green chemistry is more likely to result in loss of sensitivity when attempting to quantitate lower copy numbers.

For more information on Real-Time PCR chemistries, please refer to the following Application Notes, which you can find on our website through Technical Resources, or by entering the titles in the main Search field: “Real-Time PCR Vs. Traditional PCR”, “Essentials of Real Time PCR”, and “Selection of Reagents for Real-Time PCR”.

What are the key differences between a TaqMan MGB probe and a TaqMan TAMRA dye-labeled probe?

The TaqMan MGB probes contain the following features:
1) A fluorescent reporter at the 5' end
2) A nonfluorescent quencher at the 3' end. Because the quencher does not fluoresce, the real-time instruments can measure the reporter dye contributions more precisely.
3) A minor groove binder at the 3' end. The minor groove binder increases the melting temperature (Tm) of probes, allowing the use of shorter probes.

In general, the TaqMan MGB probes exhibit great differences in Tm values between matched and mismatched probes, which provides more accurate allelic discrimination and makes for a more sensitive real-time assay. Mismatches between a probe and allele, or target, reduce the efficiency of probe hybridization in a measurable way, which is especially important in SNP Genotyping assays. Furthermore, AmpliTaq Gold DNA polymerase is more likely to displace the mismatched probe rather than cleave it to release reporter dye. More information about TaqMan MGB probes can be found in the User Bulletin entitled "Primer Express Version 1.5 and TaqMan MGB Probes for Allelic Discrimination." You can find a copy on our website by entering this title in the main search field.

When using SYBR Green chemistry on an Applied Biosystems Real-Time PCR instrument, how do I change settings to reflect that there is no TaqMan probe being used in the reaction?

Refer to the product manual for your instrument and software for specifics, but in general you will want to change the Quencher value to None.

How can RNA standards be generated to perform absolute quantitation for RNA targets?

It is generally not possible to use DNA as a standard for absolute quantitation of RNA because there is no control for the efficiency of the reverse transcription step. Therefore, in-vitro transcribed RNA is commonly used to prepare standards for the absolute quantitation of RNA targets. This would involve the cloning of the target of interest into an in-vitro transcription plasmid, performing in-vitro transcription, then purifying the resulting cRNA so that the DNA plasmid cannot serve as a PCR template. Concentration is measured by A260 and converted to the number of copies using the molecular weight of the RNA.

Relative quantitation of gene expression methods require less up-front preparation and provide a fold-change value instead of an absolute quantity result. For many researchers, absolute quantities are not a necessary parameter to measure, and therefore relative quantitation is a much more attractive approach to studying gene expression via real-time PCR. For more information on relative quantitation of gene expression, please refer to our Technical Reference Library in the Technical Resources section of our website.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.