Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System

Catalog No. 11731023
Encompass_Preferred
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions

Includes

ThermoScript RT/Platinum Taq mix, and 2X ThermoScript reaction mix (buffer containing 0.4mM of each dNTP, 6mM MgSO4 ), vial of 50mM MgSO4

Platinum Quantitative RT-PCR ThermoScript One-Step System is for real-time quantitation of RNA molecules from total or poly(A)+ RNA in a single step.

This kit provides Platinum™ Taq DNA Polymerase, ThermoScript RT polymerase and a vial of 50mM MgSO4

Order Info

Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System contains ThermoScript™ RT/Platinum™ Taq mix, and 2X ThermoScript™ reaction mix (a buffer containing 0.4 mM of each dNTP, 6 mM MgSO4 ). A vial of 50 mM MgSO4 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Detection Method Primer-probe
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
Product Line Platinum™, ThermoScript™
Product Type RT-PCR One-Step System
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions
No. of Reactions 500 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.