Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10F' Chemically Competent E. coli

Catalog No. p-4924889
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
K203001 20 Reactions
K203040 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K203001 Supplier Invitrogen™ Supplier No. K203001
Only null left
Add to Cart
Add to Cart

Provides quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into plasmid vector

The TA Cloning Kit uses the pCR2.1 cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast and convenient: 15-minute, room-temperature ligation
  • Efficient: blue/white screening and >80% clones with correct insert
  • Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free: eliminates any enzymatic modifications of PCR product
  • Streamlined: does not require use of PCR primers that contain restriction sites
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 promoter for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

  • How TA Cloning™ Works
  • Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to 3' ends of PCR products
  • Llinearized vector supplied in this kit has single 3' deoxythymidine (T) residues
  • Allows PCR inserts to ligate efficiently with vector
  • Kit Configurations
  • TA Cloning™ Kit is offered in a variety of configurations: without competent cells (K2020-20 and K2020-40), with One Shot™ INVF' Chemically Competent E. coli (K2000-01 and K2000-40), with One Shot™ TOP10F' Chemically Competent E. coli (K2030-01 and K2030-40), and with One Shot™ TOP10 Chemically Competent E. coli (K2040-01 and K2040-40) in 20- and 40-reaction kit sizes

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10F ́
Cell Type Chemically Competent E. coli
Cloning Method TA Cloning
Content And Storage TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
For Use With (Application) PCR Cloning
No. of Reactions 20 Reactions
Product Line One Shot
Product Type Cloning Kit
Promoter T7
Quantity 20 reactions
Vector pCR2.1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.