Learn More
Invitrogen™ UltraPure™ Glycerol

Description
UltraPure Glycerol (≥99% pure) is suitable for concentrating and storing enzymes at low temperatures. It can be diluted to a 50% (v/v) solution that will not freeze at -20°C. With a density of 1.26 g/mL, it is commonly used in electrophoresis loading buffers. Additionally, UltraPure Glycerol gradients are ideal for purifying proteins, bacteriophage, and organelles.
Performance and quality testing: no DNase, RNase, or protease activity detected
Order Info
Shipping Condition: Room temperature

Specifications
Specifications
Chemical Name or Material | Glycerol |
Content And Storage | Store at room temperature. |
Product Line | UltraPure™ |
Purity | Molecular Biology Grade |
Quantity | 500 mL |
Shipping Condition | Room Temperature |
Form | Liquid |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.