Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Antibody (IVRN/2113R), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308386AF647
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey, Mouse (Negative) samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Involucrin | |
Monoclonal | |
Alexa Fluor 647 | |
involucrin | |
Recombinant human full-length Involucrin protein (IVRN) (Uniprot: P07476) | |
100 μg | |
Extracellular Matrix | |
3713 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
IVRN/2113R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Pig, Canine, Primate, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction