Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Antibody (IVRN/827), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311480PE
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human samples. It is validated for Flow CytometrySpecifications
Involucrin | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
IVRN/827 | |
Flow Cytometry | |
involucrin | |
Purified involucrin from human keratinocytes (Uniprot: P07476) | |
0.1 mL | |
Extracellular Matrix | |
3713 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction