Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Antibody (SY5), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002727MFV610
Description
Involucrin Monoclonal antibody specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Flow CytometrySpecifications
Involucrin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Pig, Canine, Primate | |
Purified |
Flow Cytometry | |
SY5 | |
Flow Cytometry | |
involucrin | |
Purified involucrin from human keratinocytes. | |
0.1 mL | |
Extracellular Matrix | |
3713 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction