Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Antibody (SY8), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002774AF647
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Involucrin | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IVL | |
Purified involucrin from human keratinocytes. | |
0.1 mL | |
Extracellular Matrix | |
3713 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SY8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
involucrin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Pig, Canine, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction