Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Antibody (SY5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002727JF549
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Involucrin | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IVL | |
Purified involucrin from human keratinocytes. | |
0.1 mL | |
Primary | |
Canine, Human, Pig, Primate | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SY5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
involucrin | |
Mouse | |
Protein A purified | |
Extracellular Matrix | |
3713 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction