Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Mouse anti-Human, Porcine, Canine, Primate, Monkey, Clone: rIVRN/827, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Involucrin |
---|---|
Clone | rIVRN/827 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Involucrin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Extracellular Matrix | |
10 mM PBS with 0.05% BSA | |
3713 | |
IgG1 κ | |
Protein A or G purified |
rIVRN/827 | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Pig, Canine, Primate, Monkey | |
involucrin | |
Recombinant human full-length Involucrin protein (IVRN) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title