Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INVS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121821
This item is not returnable.
View return policy
Description
INVS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein containing multiple ankyrin domains and two IQ calmodulin-binding domains. The encoded protein may function in renal tubular development and function, and in left-right axis determination. This protein interacts with nephrocystin and infers a connection between primary cilia function and left-right axis determination. A similar protein in mice interacts with calmodulin. Mutations in this gene have been associated with nephronophthisis type 2. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
INVS | |
Polyclonal | |
Unconjugated | |
Invs | |
INV; inversin; Inversion of embryo turning homolog; inversion of embryo turning protein; inversion of embryonic turning; INVS; nephrocystin 2; nephrocystin-2; NPH2; NPHP2; RGD1563359 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27130 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y283 | |
Invs | |
Recombinant protein Nephronophthisis 2, Infantile. The antigen corresponds to amino acid range 1-237 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction