Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodic acid, 99%

Catalog No. p-7081777
Click to view available options
Quantity:
50 g
250 g
1000 g

Catalog No. AAA1192518

Supplier: Thermo Scientific Chemicals A1192518

Only null left
Add to Cart

CAS: 7782-68-5 | ILiO3 | 181.84 g/mol

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

Solubility
Soluble in water and methanol.

Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Specifications

Melting Point 110°C (decomposition)
Density 4.629
Odor Odorless
Sensitivity Light sensitive
Quantity 50 g
UN Number UN3085
Merck Index 14,5012
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
Solubility Information Soluble in water and methanol.
Formula Weight 175.91
Percent Purity 99%
Chemical Name or Material Iodic acid
Show More Show Less
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber 231-962-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.