Learn More
Iodic acid, 99%
Description
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.

Specifications
Specifications
Melting Point | 110°C (decomposition) |
Density | 4.629 |
Odor | Odorless |
Sensitivity | Light sensitive |
Quantity | 1000 g |
UN Number | UN3085 |
Merck Index | 14,5012 |
Synonym | unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 175.91 |
Show More |
Safety and Handling
Hazard Category | H272-H314-H335 |
Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S. |
EINECSNumber | 231-962-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.