Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodide Standard Solution, 0.100M, Fisher Chemical™
SDP

Catalog No. SI90011 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 7681-11-0, 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

CAS Min % 96.0, 1.6
CAS Max % 100.0, 1.67
Grade Laboratory
Packaging Amber Glass Bottle
Methods For AOAC Method 992.22
Quantity 1 L
Chemical Name or Material Iodide Standard Solution, 0.100M
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.