Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, 0.0100 Normal (N/100), Reagents

Catalog No. 01334942
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-942 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-942 Supplier Reagents Holdings Llc Supplier No. CI1009001C
Only null left
Add to Cart
Add to Cart

Iodine, 0.0100 Normal (N/100), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Brown
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0051
Chemical Name or Material Iodine, 0.0100 Normal (N/100)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.