Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, 0.08 Normal in Methanol, Reagents
Click to view available options
Quantity:
1 L
4 L
Specifications
Specifications
| Boiling Point | 64.5°C |
| Color | Dark Brown |
| Physical Form | Liquid |
| Quantity | 4 L |
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 0.8 |
| Chemical Name or Material | Iodine, 0.08 Normal in Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction