Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, 0.1 N Solution Volumetric Solution, USP, Spectrum™ Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Iodine, 0.1 N Solution, USP Volumetric Solution is a ready-to-use chemical solution used in the analysis of many finished USP grade products. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1.3%, 4% |
Concentration | 0.09500-0.10500 N |
Grade | Chemical Solution |
Packaging | Amber Glass Bottle |
Quantity | 4 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction