Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, D'Antoni, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 0.96, 96.0, 0.96 |
CAS Max % | 1.0, 100.0, 1.0 |
Grade | Laboratory |
Packaging | Amber Glass Bottle |
Color | Dark Brown |
Quantity | 1 L |
Physical Form | Liquid |
Chemical Name or Material | Iodine, D'Antoni |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction