Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monochloride, 1M soln. in dichloromethane

Catalog No. p-7031014
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7790-99-0 | ClI | 162.35 g/mol

Iodine monochloride is a useful reagent in organic synthesis and source of I+. It is used as a source of electrophilic iodine in the synthesis of certain aromatic iodides. It also cleaves C-Si bonds. Iodine monochloride will also add to the double bond in alkenes to give chloro-iodo alkanes. It can be dissolved in acetic acid to make Wijs solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Linear Formula ICl
UN Number UN2922
Concentration 1M soln. in dichloromethane
Density 1.42
Solubility Information Insoluble in water.
Formula Weight 162.36
Quantity 100 mL
Sensitivity Air and light sensitive
Chemical Name or Material Iodine monochloride
Hazard Category H300-H314-H335-H336-H350
Hazard Statement GHS H Statement
H314-H351
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-236-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.