Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monochloride, 1M solution in dichloromethane, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
Glass bottle
7790-99-0,75-09-2
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride,iodine chloride,iodochlorine,iodine chloride icl,chloroiodide,wijs' chloride,iodinemonochloride,chlorine iodide,unii-0smg5nlu45,protochlorure d'iode french
24640
ClI
7790-99-0,75-09-2
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride,iodine chloride,iodochlorine,iodine chloride icl,chloroiodide,wijs' chloride,iodinemonochloride,chlorine iodide,unii-0smg5nlu45,protochlorure d'iode french
24640
ClI
C-X bond formation reagent
Linear Formula ICl
Fieser 01,502; 08,261; 10,212; 11,268
Merck Index 15, 5060
Solubility Information Solubility in water: insoluble
Formula Weight 162.35
Physical Form Solution
Concentration 0.95 to 1.05M
Density 1.4200g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Specific Gravity 1.42
Color Brown to Orange
Quantity 800mL
Chemical Name or Material Iodine monochloride, 1, 0M solution in dichloromethane
Show More Show Less

GHS H Statement
Suspected of causing cancer.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
R

GHS Signal Word: Danger

EINECSNumber : 232-236-7