Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monochloride, ACS

Catalog No. p-4407450
Click to view available options
Quantity:
25 g
100 g
500 g

Moisture and Light Sensitive | CAS: 7790-99-0 | ClI | 162.35 g/mol

Iodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point 96°C to 98°C
Melting Point 25°C to 27°C
Color Black-Red
Physical Form Crystalline, Liquid
Quantity 25 g
Beilstein 3902972
Solubility Information Soluble in water,alcohol,ether,carbon disulfide.
Formula Weight 162.36
Grade ACS
Sensitivity Light and moisture sensitive
Appearance Red-black crystalline/liquid
Density 3.182
Chemical Name or Material Iodine monochloride, For analysis ACS
Show More Show Less
Hazard Category H300-H311-H314-H335-H373
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-236-7
RTECSNumber NN1650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.