Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Monochloride, Solution, Spectrum™ Chemical
SDP

Catalog No. 18611065 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle

Specifications

Chemical Name or Material Iodine Monochloride
Color Clear, Light Brown
CAS 7790-99-0, 64-19-7
CAS Max % 1.68, 98.32
Grade Chemical Solution
Packaging Amber Glass Bottle
Suitability To Pass Test
Quantity 4 L

CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.