Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution, 5%, Lugol, Fisher Chemical

Catalog No. si102500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2, 7681-11-0, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown
Boiling Point 100°C
Density 1.4g/cm³
Percent Purity 4.5 to 5.5%
Identification Pass test
Packaging Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Iodine Solution, 5%, Lugol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.