Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution, Wijs, Fisher Chemical™

Catalog No. SI1061 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SI106-1 1 L Amber Glass
SI106-500 500 mL Amber Glass
SI106-4 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SI106-1
Add to Cart

For determination of iodine absorption number in fats. Approximately 0.22N solution in glacial acetic acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2 , 64-19-7 , 7782-50-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Amber
Boiling Point 118°C
Melting Point 17°C
UN Number 2920
Concentration 0.215 to 0.225
Density 1.06g/cm³
Identification Pass test
Packaging Amber Glass
Vapor Pressure 11mmHg at 20°C
pH 2.0
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Iodine Solution, Wijs
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.