Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Iodine, Stabilized
SDP

Catalog No. p-200005337 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Includes:
250 mL
3.7 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-001-814 250 mL
23-001-771 3.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-814 Supplier Hardy Diagnostics™ Supplier No. I008
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A mordant used in the gram stain procedure. A stabilized, thicker iodine solution

  • Stabilized Iodine is thicker and requires a longer decolorization step or a faster decolorizer
  • Stabilized Iodine is less sensitive to light and has a longer shelf life

Specifications

Includes 250 mL
Format Liquid
Color Dark Amber to Brown
Boiling Point 184°C
Storage Requirements Room Temperature
For Use With (Equipment) Gram Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.