Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18603483 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 25 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

I1010, 7553-56-2, I2

Spectrum™ Chemical Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS is a chemical element and an analytical reagent. As an ACS grade Reagent, Spectrum Chemical manufactured Iodine is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity. This item is a DEA List I Chemical and does require the completion of a Regulatory form.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 100%
Quantity 6 x 125 g
UN Number UN3495
Percent Purity 99.8%
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.