Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18603488 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 25 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-488 12 kg Poly Pail
18-603-480 25 g Amber Glass Bottle
18-603-482 125 g Amber Glass Bottle
18-603-481 6 x 25 g Amber Glass Bottle
18-603-484 500 g Amber Glass Bottle
18-603-483 6 x 125 g Amber Glass Bottle
18-603-486 2.5 kg Amber Glass Bottle
18-603-485 6 x 500 g Amber Glass Bottle
18-603-487 4 x 2.5 kg Amber Glass Bottle
18-603-489 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18-603-488 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I101012KG
Add to Cart
Add to Cart
Request a Quote

I1010, 7553-56-2, I2

Spectrum™ Chemical Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS is a chemical element and an analytical reagent. As an ACS grade Reagent, Spectrum Chemical manufactured Iodine is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity. This item is a DEA List I Chemical and does require the completion of a Regulatory form.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 100%
Quantity 12 kg
UN Number UN3495
Percent Purity 99.8%
Grade ACS
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.