Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Tincture, approx. 2% Solution
SCI_ED

Catalog No. s25944 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Liquid

Chemical Identifiers

CAS 7553-56-2, 64-17-5, 7681-82-5, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown
Physical Form Liquid
Quantity 500 mL
Grade Laboratory
Packaging Liquid
Density 0.900
Chemical Name or Material Iodine, Tincture, approx. 2% Solution
Recommended Storage Stable under normal conditions of use and storage
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.