Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine trichloride, 97%

Catalog No. p-7037297
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 865-44-1 | Cl3I | 233.254 g/mol

Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

Solubility
Soluble with decomposition.

Notes
Light Sensitive. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-44-1
Molecular Formula Cl3I
Molecular Weight (g/mol) 233.254
MDL Number MFCD00036291
InChI Key PAWIVBWALDNUJP-UHFFFAOYSA-N
Synonym iodine trichloride, iodine chloride icl3, unii-1e5kq66trq, 1e5kq66trq, jodtrichlorid, trichloro-, e3-iodane, trichloro-$l^ 3-iodane, ksc450m5f, iodine trichloride, saj first grade
PubChem CID 70076
IUPAC Name trichloro-$l^{3}-iodane
SMILES ClI(Cl)Cl

Specifications

Quantity 5 g
Linear Formula ICl3
UN Number UN3085
Merck Index 14,5020
Solubility Information Soluble with decomposition.
Formula Weight 233.26
Percent Purity 97%
Sensitivity Light sensitive
Chemical Name or Material Iodine trichloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 212-739-8
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.