Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine(V) oxide, 98%
Click to view available options
Quantity:
25 g
100 g
Description
Iodine (V) oxide is used as an oxidizing agent to remove carbon monoxide from gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 275°C to 350°C (decomposition) |
| Color | White to Cream |
| pH | 1.1 |
| Physical Form | Crystalline |
| Quantity | 100 g |
| UN Number | UN3085 |
| Merck Index | 14,5019 |
| Solubility Information | Soluble in water and nitric acid; Insoluble in absolute alcohol,chloroform,ethanol,ether and carbon disulfide. |
| Formula Weight | 333.81 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H272-H314 |
| Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: III |
| EINECSNumber | 234-740-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction