Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine(V) oxide, 98%

Catalog No. p-7080376
Click to view available options
Quantity:
25 g
100 g

An oxidizing agent to remove CO from gas | CAS: 12029-98-0 | I2O5 | 333.804 g/mol

Iodine (V) oxide is used as an oxidizing agent to remove carbon monoxide from gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12029-98-0
Molecular Formula I2O5
Molecular Weight (g/mol) 333.804
MDL Number MFCD00011356
InChI Key BIZCJSDBWZTASZ-UHFFFAOYSA-N
Synonym iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide
PubChem CID 159402
ChEBI CHEBI:29914
IUPAC Name iodyl iodate
SMILES O=I(=O)OI(=O)=O

Specifications

Melting Point 275°C to 350°C (decomposition)
Color White to Cream
pH 1.1
Physical Form Crystalline
Quantity 25 g
UN Number UN3085
Merck Index 14,5019
Solubility Information Soluble in water and nitric acid; Insoluble in absolute alcohol,chloroform,ethanol,ether and carbon disulfide.
Formula Weight 333.81
Percent Purity 98%
Sensitivity Hygroscopic; Light sensitive
Density 5.08 g/mL
Chemical Name or Material Iodine (V) oxide
Show More Show Less
Hazard Category H272-H314
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 234-740-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.