Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT) PROMO
Click to view available options
:
Thermo Scientific™ Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT)
Description
- A sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis
- Provided in convenient single-use, no-weigh format
- High-purity, crystalline reagent
- Compatible with EasyPep chemistry and High Select Phosphopeptide Enrichment kits
Specifications
Specifications
Quantity | 10 x 3.5 mg |
Molecular Weight (g/mol) | 348.99 |
Percent Purity | >90% |
Grade | MS |
Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction