Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoacetic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
50g
100g
Packaging:
Glass bottle
64-69-7
C2H3IO2
185.95
MFCD00002685
JDNTWHVOXJZDSN-UHFFFAOYSA-N
iodoacetic acid,acetic acid, iodo,iodoacetate,monoiodoacetic acid,monoiodoacetate,monoiodine acetate,iodoaceticacid,iodoethanoic acid,acetic acid, 2-iodo,kyselina jodoctova
5240
CHEBI:74571
2-iodoacetic acid
C(C(=O)O)I
64-69-7
C2H3IO2
185.95
MFCD00002685
JDNTWHVOXJZDSN-UHFFFAOYSA-N
iodoacetic acid,acetic acid, iodo,iodoacetate,monoiodoacetic acid,monoiodoacetate,monoiodine acetate,iodoaceticacid,iodoethanoic acid,acetic acid, 2-iodo,kyselina jodoctova
5240
CHEBI:74571
2-iodoacetic acid
C(C(=O)O)I
Melting Point 79.0°C to 83.0°C
Infrared Spectrum Authentic
Color Brown to Yellow
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula ICH2CO2H
Beilstein 02, 222
Merck Index 15, 5068
Solubility Information Solubility in water: 600g/L (20 c). Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 185.95
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Iodoacetic acid, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 200-590-1

RTECSNumber : AI3500000

TSCA : TSCA