Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene, 98%

Catalog No. p-7020731
Click to view available options
Quantity:
50 g
100 g
1000 g

Catalog No. AAA1278818

Supplier: Thermo Scientific Chemicals A1278818

Only null left
Add to Cart

CAS: 591-50-4 | C6H5I | 204.01 g/mol

Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.

Solubility
Miscible with ethanol, chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.

Notes
Light sensitive. Incompatible with magnesium and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point °C to 31°C
Density 1.826
Boiling Point 188°C to 189°C
Flash Point 74°C (165°F)
Assay Percent Range 98%
Refractive Index 1.62
Quantity 50 g
Beilstein 1446140
Sensitivity Light sensitive
Merck Index 14,5029
Solubility Information Miscible with ethanol,chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.
Formula Weight 204.01
Percent Purity 98%
Chemical Name or Material Iodobenzene
Show More Show Less
Hazard Category H227-H302+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
EINECSNumber 209-719-6
RTECSNumber DA3390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.