Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
591-50-4
C6H5I
204.01
SNHMUERNLJLMHN-UHFFFAOYSA-N
benzene, iodo,phenyl iodide,benzene iodide,iodophenyl,iodinebenzol,iodo-benzene,phenyliodide,iodo benzene,unii-9hk5l7ybbr,1-iodobenzene
11575
iodobenzene
C1=CC=C(C=C1)I
591-50-4
C6H5I
204.01
SNHMUERNLJLMHN-UHFFFAOYSA-N
benzene, iodo,phenyl iodide,benzene iodide,iodophenyl,iodinebenzol,iodo-benzene,phenyliodide,iodo benzene,unii-9hk5l7ybbr,1-iodobenzene
11575
iodobenzene
C1=CC=C(C=C1)I
Density 1.8200g/mL
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.6180 to 1.6200
Specific Gravity 1.82
Quantity 500mL
Flash Point 74°C
Melting Point -29.0°C
Boiling Point 188.0°C
Assay Percent Range 98%
Linear Formula C6H5I
Beilstein ,215
Fieser 01,505
Merck Index 15,5072
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Formula Weight 204.01
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Iodobenzene
Show More Show Less