Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene, 98%

Manufacturer:  Alfa Aesar A1278818

Quantity 50g

 View more versions of this product

Catalog No. AAA1278818


  Check Availability
Add to cart

Specifications

Description

Applications
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.

Solubility
Miscible with ethanol, chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.

Notes
Light sensitive. Incompatible with magnesium and strong oxidizing agents.
Description & Specifications

Chemical Identifiers

591-50-4
204.01
1446140
Iodobenzene
C6H5I
MFCD00001029
14,5029

Specifications

591-50-4
1.826
204.01
1446140
50g
98%
98%
C6H5I
MFCD00001029
14,5029
Iodobenzene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P261-P280f
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.

Warning

RTECSNumber : DA3390000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit