Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoethane, 98+%, stab. with copper

Catalog No. p-7021912
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 75-03-6 | C2H5I | 155.966 g/mol

Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-03-6
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.966
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
PubChem CID 6340
ChEBI CHEBI:42487
IUPAC Name iodoethane
SMILES CCI

Specifications

Melting Point -108°C
Density 1.95
Boiling Point 70°C to 73°C
Flash Point >73°C (163°F)
Refractive Index 1.512
Quantity 100 g
Beilstein 505934
Sensitivity Light and moisture sensitive
Merck Index 14,3813
Solubility Information Miscible with water.
Formula Weight 155.97
Percent Purity ≥98%
Chemical Name or Material Iodoethane, stabilized with copper
Show More Show Less
Hazard Category H227-H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
EINECSNumber 200-833-1
RTECSNumber KI4750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.