Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
74-88-4,1634-04-4
CH3I
141.94
MFCD00001073
INQOMBQAUSQDDS-UHFFFAOYSA-N
methyl iodide,monoiodomethane,methane, iodo,methyljodid,jod-methan,methyliodide,methyljodide,iodometano,joodmethaan,metylu jodek
6328
CHEBI:39282
iodomethane
CI
74-88-4,1634-04-4
CH3I
141.94
MFCD00001073
INQOMBQAUSQDDS-UHFFFAOYSA-N
methyl iodide,monoiodomethane,methane, iodo,methyljodid,jod-methan,methyliodide,methyljodide,iodometano,joodmethaan,metylu jodek
6328
CHEBI:39282
iodomethane
CI
Alkylation reagent
Appearance Clear colorless, may gain color on storage
Density 0.9330g/mL
Quantity 800mL
Flash Point −18°C
Boiling Point 41.0°C to 43.0°C
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.933
Merck Index 15, 6159
Formula Weight 141.94
Chemical Name or Material Iodomethane, 2M Solution in tert-Butyl Ether

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/spark

GHS Signal Word: Danger

EINECSNumber : 200-819-5