Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethane, 99+%, stab. with copper

Catalog No. p-3257069
Click to view available options
Quantity:
100 g
500 g

CAS: 74-88-4 | CH3I | 141.94 g/mol

Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
MDL Number MFCD00001073
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
Synonym methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek
PubChem CID 6328
ChEBI CHEBI:39282
IUPAC Name iodomethane
SMILES CI

Specifications

Melting Point -67°C
Density 2.28
Boiling Point 41°C to 43°C
Assay Percent Range 99+%
Refractive Index 1.53
Quantity 100 g
Beilstein 969135
Sensitivity Light sensitive
Solubility Information Miscible with water.
Formula Weight 141.94
Physical Form Liquid
Chemical Name or Material Iodomethane
Show More Show Less
Hazard Category H301+H331-H312-H315-H319-H335-H351
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 200-819-5
RTECSNumber PA9450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.