Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethane, Stabilized, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
74-88-4,7440-50-8
CH3I
141.94
INQOMBQAUSQDDS-UHFFFAOYSA-N
methyl iodide,monoiodomethane,methane, iodo,methyljodid,jod-methan,methyliodide,methyljodide,iodometano,joodmethaan,metylu jodek
6328
CHEBI:39282
iodomethane
CI
74-88-4,7440-50-8
CH3I
141.94
INQOMBQAUSQDDS-UHFFFAOYSA-N
methyl iodide,monoiodomethane,methane, iodo,methyljodid,jod-methan,methyliodide,methyljodide,iodometano,joodmethaan,metylu jodek
6328
CHEBI:39282
iodomethane
CI
Alkylation reagent
Color Undesignated
Viscosity 0.5 mPa.s (20°C)
Specific Gravity 2.28
Melting Point -66.0°C
Infrared Spectrum Authentic
Refractive Index 1.5290 to 1.5310
Stabilizer Copper
Water 0.1% max.
Boiling Point 42.5°C
Packaging Glass bottle
Density 2.2800g/mL
Quantity 2.5kg
Assay Percent Range 99%
Linear Formula CH3I
Beilstein 01, 69
Fieser 01,682; 02,274; 03,202; 04,341; 05,447; 06,384; 08,342; 09,308; 15,118
Merck Index 15, 6159
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide
Formula Weight 141.94
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Iodomethane
Show More Show Less

GHS H Statement
Toxic if swallowed. Harmful in contact with skin. Causes skin irritation. Toxic if inhaled. May cause respiratory irritation. Suspected of causing cancer.

GHS P Statement
Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.

Danger