Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethylcyclopentane, 98%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
27935-87-1
C6H11I
210.05
DUMSKQUKLVSSII-UHFFFAOYSA-N
iodomethyl cyclopentane,cyclopentylmethyl iodide,cyclopentane, iodomethyl,cyclopentyliodomethane,acmc-20akpz,cyclopentylmethyliodide,iodomethyl-cyclopentane,cyclopentanemethyl iodide
2762531
iodomethylcyclopentane
C1CCC(C1)CI
27935-87-1
C6H11I
210.05
DUMSKQUKLVSSII-UHFFFAOYSA-N
iodomethyl cyclopentane,cyclopentylmethyl iodide,cyclopentane, iodomethyl,cyclopentyliodomethane,acmc-20akpz,cyclopentylmethyliodide,iodomethyl-cyclopentane,cyclopentanemethyl iodide
2762531
iodomethylcyclopentane
C1CCC(C1)CI
Additional Information Stabilized over copper
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H9)CH2I
Solubility Information Solubility in water: immiscible
Formula Weight 210.05
Physical Form Solution
Percent Purity 98%
Density 1.6000g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5400 to 1.5440
Solubility Solubility in water: immiscible.
Specific Gravity 1.6
Stabilizer copper powder
Storage Information May darken on storage
Boiling Point 78.0°C (17.0 mmHg)
Color Brown or Colorless to Orange
Quantity 1g
Flash Point 92°C
Chemical Name or Material Iodomethylcyclopentane, Stabilized
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning